missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Magnetic Plastic Transport Signs
£20.80 - £21.80
Specifications
Material | Plastic |
---|---|
Quantity | 1 Ea. |
Adhesive Type | Permanent Acrylic |
Description | Magnetic Plastic |
Product Type | Transport Sign |
Product Code | Brand | Displayed Text | Shape | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Shape | Price | Quantity & Availability | ||||
11774914
|
Brady™
258976 |
Corrosive Substance 8 | Diamond |
£21.80
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
11794914
|
Brady™
258979 |
Miscellaneous hazardous substances 9 | Diamond |
£21.80
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
11714914
|
Brady™
258961 |
Oxidizing Substance 5.1 | Diamond |
£21.80
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
11734914
|
Brady™
258967 |
Toxic and Harmful Substances 6 | Diamond |
£20.80
Each |
In Stock
Log in to see stock. |
||||
11721003
|
Brady™
813603 |
Environmentally Hazardous Substances | Rhomboid |
£21.35
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
11714904
|
Brady™
258943 |
Non-flammable, non-toxic gas 2 | Rhomboid |
£21.80
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
11730244
|
Brady™
223599 |
Non-flammable, non-toxic gas 2 | Diamond |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11750953
|
Brady™
810938 |
Organic Peroxide 5.2 | Diamond |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11700963
|
Brady™
810944 |
Organic Peroxide 5.2 | Rhomboid |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Plastic | |
Permanent Acrylic | |
Transport Sign |
1 Ea. | |
Magnetic Plastic |