missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog Number p-3257037
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

For Fireproofing, Textiles, Catalyst Carrier

Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in in water and acid. Slightly soluble in alcohol and glycerol.
Formula Weight 246.48 (120.37 Anhydrous)
Grade ACS
Odor Odorless
Appearance White crystals
Density 1.68 g/cm3 at 20°C
Chemical Name or Material Magnesium sulfate heptahydrate, heptahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'