missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Extra Pure, SLR, Dried, Fisher Chemical™

Catalog Number 10776101
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


32.97 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
500 g
1 kg
2 kg
10 kg
Packaging:
Plastic keg
Plastic powder jar
22189-08-8
MgO4S
120.36
MFCD00149787
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
[Mg++].[O-]S([O-])(=O)=O
22189-08-8
MgO4S
120.36
MFCD00149787
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
[Mg++].[O-]S([O-])(=O)=O

Magnesium Sulfate, Extra Pure, SLR, Dried

TRUSTED_SUSTAINABILITY
Melting Point 1124°C
Color White
Physical Form Powder/Solid
Quantity 1 kg
IUPAC Name magnesium(2+) sulfate
Formula Weight 120.36g/mol
Packaging Plastic powder jar
Vapor Pressure <0.1mmHg at 20°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'