missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 7439-95-4 |
Molecular Formula | Mg |
MDL Number | MFCD00085308 |
Quantity | 1 Ea. |
Synonym | magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs |
InChI Key | JLVVSXFLKOJNIY-UHFFFAOYSA-N |
SMILES | [Mg] |
IUPAC Name | magnesium |
Molecular Weight (g/mol) | 24.30 |
PubChem CID | 5462224 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'