missing translation for 'onlineSavingsMsg'
Learn More

Magnesium silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407622
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
22831-39-6
Mg2Si
76.695
MFCD00016202
YTHCQFKNFVSQBC-UHFFFAOYSA-N
magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
89858
[Mg]=[Si]=[Mg]
This item is not returnable. View return policy
22831-39-6
Mg2Si
76.695
MFCD00016202
YTHCQFKNFVSQBC-UHFFFAOYSA-N
magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
89858
[Mg]=[Si]=[Mg]

Air and Moisture Sensitive; Decomposes in water, HCl

it is used as an intermediate in chemical research,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1102°C
Color Blue
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Insoluble in water and denser than water.
Formula Weight 76.71
Sensitivity Air and moisture sensitive
Appearance Blue powder
Density 2
Chemical Name or Material Magnesium silicide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'