missing translation for 'onlineSavingsMsg'
Learn More

Magnesium phosphate pentahydrate, P{2}O{5} 33%, Thermo Scientific Chemicals

Catalog Number 11303437
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
10233-87-1
H10Mg3O13P2
352.93
MFCD00150252
QZYXGKYJDBUYES-UHFFFAOYSA-H
magnesium phosphate pentahydrate, magnesium phosphate usp, phosphoric acid, magnesium salt 2:3 , pentahydrate, 3mg.2po4.5h2o, trimagnesium phosphate pentahydrate, magnesium phosphate 3:2 pentahydrate, magnesium phosphate, tribasic pentahydrate, magnesium phosphate pentahydrate, p2o, trimagnesium 2+ pentahydrate diphosphate
23724945
trimagnesium;diphosphate;pentahydrate
O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Mg+2].[Mg+2].[Mg+2]
This item is not returnable. View return policy
10233-87-1
H10Mg3O13P2
352.93
MFCD00150252
QZYXGKYJDBUYES-UHFFFAOYSA-H
magnesium phosphate pentahydrate, magnesium phosphate usp, phosphoric acid, magnesium salt 2:3 , pentahydrate, 3mg.2po4.5h2o, trimagnesium phosphate pentahydrate, magnesium phosphate 3:2 pentahydrate, magnesium phosphate, tribasic pentahydrate, magnesium phosphate pentahydrate, p2o, trimagnesium 2+ pentahydrate diphosphate
23724945
trimagnesium;diphosphate;pentahydrate
O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Mg+2].[Mg+2].[Mg+2]

Magnesium phosphate pentahydrate is used in the preparation of magnesium calcium phosphate biocement(MCPB) for bone regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystals
Quantity 1 kg
Assay Percent Range P2O5 33%
Solubility Information Insoluble in water.
Formula Weight 352.93 (262.86 Anhydrous)
Appearance White crystals
Density g/cm3 at 20°C
Chemical Name or Material Magnesium phosphate pentahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'