missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate, Pure, Dried, Desiccating Agent, Fisher Chemical™

Brand:  Fisher Chemical M/0950/53

Code : 21

Additional Details : CAS Number : 10034-81-8 Weight : 0.50000kg Transport : UN number : 1475 Chem class : 5.1 Pack group : II


Quantity 500g

 View more versions of this product

Product Code. 10050240

Quantity Price
1 £364.36 / 500g
10 £315.96 / 500g
Estimated Shipment: 02-02-2022

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Chemical Identifiers

10034-81-8
223.197
MPCRDALPQLDDFX-UHFFFAOYSA-L
24840
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
Cl2MgO8
11102
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
magnesium;diperchlorate

Specifications

10034-81-8
11102
MPCRDALPQLDDFX-UHFFFAOYSA-L
magnesium;diperchlorate
24840
Solid
White
5 to 9
Cl2MgO8
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
223.197
223.21g/mol
Plastic powder jar
250°C
500g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Magnesium perchlorate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates
Special Offers

Special Offers