missing translation for 'onlineSavingsMsg'
Learn More

Magnesium perchlorate, ACS, Thermo Scientific Chemicals

Catalog Number p-4407613
Click to view available options
Quantity:
100 g
500 g
10034-81-8
Cl2MgO8
223.197
MFCD00011102
MPCRDALPQLDDFX-UHFFFAOYSA-L
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
24840
magnesium;diperchlorate
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
10034-81-8
Cl2MgO8
223.197
MFCD00011102
MPCRDALPQLDDFX-UHFFFAOYSA-L
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
24840
magnesium;diperchlorate
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Magnesium Perchlorate, Anhydrous, Cl2MgO8, CAS Number-10034-81-8, magnesium perchlorate, anhydrous, anhydrous magnesium perchlorate, anhydrone, magnesium perchlorate, magnesium diperchlorate, hsdb 661, dehydrite, perchlorate de magnesium french, perchloric acid, magnesium salt, unii-7n77z541yf, 500g

Magnesium perchlorate is commonly utilized as a potent oxidizing agent. It is used as dehydrating agent in organic synthesis. It is practiced in many domains like pharma industry, pyrotechnics industry,in rechargeable batteries and it is a versatile tool in organic synthesis. It possesses photoelectronic and optical properties. Thus it can be used in photo electronic and optoelectronic devices. It is applied as a strong drying agent for gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 251°C (decomposition)
Physical Form Granules
Quantity 100 g
Beilstein EM>5,
Solubility Information Magnesium perchlorate is soluble in water,ethanol.
Formula Weight 223.21
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.21 g/mL
Chemical Name or Material Magnesium perchlorate, anhydrous, For analysis ACS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'