Magnesium oxide, White powder, 96% min., Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 044078.36

MAGNESIUM OXIDE 96% MIN 500G

UNSPSC: 12141502

Code: NEW

Additional Details:
Additional Details: CAS Number: 1309-48-4 Weight: 0.01000kg


Quantity 500g

Product Code. 11305168

Quantity Price
1 £ 100.17 / 500g
Estimated Shipment date
from Supplier 07-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Magnesium oxide
CAS 1309-48-4
Physical Form Powder
Assay Percent Range 96% min.
Molecular Formula MgO
Formula Weight 40.3
MDL Number MFCD00011109

Specification

Physical Form Powder
Formula Weight 40.3
Quantity 500g
Color White
Chemical Name or Material Magnesium oxide
Assay Percent Range 96% min.
Molecular Formula MgO
MDL Number MFCD00011109
CAS 1309-48-4