missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 96% min, Thermo Scientific Chemicals

Catalog Number p-4824800
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]

Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, a protective coating in plasma displays, an oxide barrier in spin-tunneling devices, an optical material, a component of tubular construction in heating elements, and also in libraries for preserving books. Magnesium oxide microspheres used in enrichment method of polycyclic aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 3600°C
Melting Point 2852°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min.
Solubility Information Soluble in acid and ammonia. Insoluble in water and ethanol.
Formula Weight 40.3
Refractive Index 1.736
Density 3.58
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'