missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis excluding Ca), Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 1 Ea. |
MDL Number | MFCD00011109 |
Formula Weight | 40.3 |
Percent Purity | 99.95% |
Assay | (metals basis excluding Ca) |
Chemical Name or Material | Magnesium oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'