Alfa Aesar™ Magnesium oxide, 99.99% (metals basis)


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 043186.36

UNSPSC: 12141502

Code: NEW

Additional Details:
Additional Details: CAS Number: 1309-48-4 Weight: 0.01000kg


Quantity 500g

Product Code. 11304888

Quantity Price
1 £ 1554.81 / 500g
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Magnesium oxide
CAS 1309-48-4
Molecular Formula MgO
Formula Weight 40.3
MDL Number MFCD00011109
Merck Index 14,5677

Specification

Formula Weight 40.3
TSCA TSCA
Quantity 500g
Physical Form Powder
Refractive Index 1.736
Density 3.58g/mL
Melting Point 2852°C
Molecular Formula MgO
CAS 1309-48-4
Boiling Point 3600°C
Assay Percent Notes (metals basis)
Odor Odorless
Sensitivity Air Sensitive
MDL Number MFCD00011109
Chemical Name or Material Magnesium oxide
Merck Index 14,5677
Assay 99.99%