missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 95% min, Thermo Scientific Chemicals

Catalog Number p-7058761
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]

For plasma spraying, Refractory applications, electrical insulation, pharmaceuticals, cosmetics, paper manufacture, adsorption and catalysis, and in semiconductors, reflector in optical instruments. Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, a protective coating in plasma displays, an oxide barrier in spin-tunneling devices, an optical material, a component of tubular construction in heating elements, and also in libraries for preserving books. Magnesium oxide microspheres used in enrichment method of polycyclic aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For plasma spraying, Refractory applications, electrical insulation, pharmaceuticals, cosmetics, paper manufacture, adsorption and catalysis, and in semiconductors, reflector in optical instruments
TRUSTED_SUSTAINABILITY
Boiling Point 3600°C
Melting Point 2852°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 95% min
Merck Index 14,5677
Solubility Information Slightly soluble in water,(solubility increased by CO2); Soluble in acids and ammonium salt solution; Insoluble in alcoholSoluble in acid and ammonia; Insoluble in water and ethanol.
Formula Weight 40.3
Percent Purity ≥95%
Refractive Index 1.736
Density 3.58
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'