missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number p-5547819
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]

Magnesium oxide is used as a refractive material, which is physically and chemically stable at higher temperature. Magnesium oxide is a raw material for producing portland cement. It is also used in agricultural, chemical construction, environment al and other industrial applications. It is also used in medicine, colorimetry, industrial cables, heating elements, ceramics, spin-tunneling devices, and several other materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 3600°C
Melting Point 2852°C
Color White
Physical Form Fused Lump
Quantity 25 g
Assay Percent Range 99.95% (Metals basis)
Solubility Information Soluble in acid and ammonia. Insoluble in alcohol.
Formula Weight 40.3
Refractive Index 1.736
Density 3.58 g/cm3 at 20°C
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'