missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Magnesium nitrate hydrate, Puratronic™, 99.999% (metals basis)

Brand:  Alfa Aesar™ 010799.22

Code : NEW

Additional Details : CAS Number : 10377-60-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11306907

  • £222.70 / 100g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Magnesium nitrate hydrate
Puratronic™
MFCD00011103
14,5674
10377-60-3
148.32 (Anhydrous)
UN1474

Specifications

Magnesium nitrate hydrate
Puratronic™
99.999%
148.32 (Anhydrous)
UN1474
1.46g/mL
Hygroscopic
89°C
10377-60-3
Crystalline Aggregates
(metals basis)
MFCD00011103
14,5674
Odorless
330°C (dec.)
100g
Videos
SDS