missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium nitrate hexahydrate, 99.97% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
250 g
Description
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Boiling Point | 330°C (decomposition) |
Melting Point | 89°C |
Physical Form | Crystalline |
Quantity | 5 g |
Assay Percent Range | 99.97% (metals basis) |
Solubility Information | Very soluble in water,alcohol and liquid ammonia. |
Formula Weight | 256.41 (148.32 Anhydrous) |
Sensitivity | Hygroscopic |
Density | 1.46 g/mL |
Chemical Name or Material | Magnesium nitrate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'