missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-44476
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
13446-18-9
H12MgN2O12
256.40
MFCD00149779
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
202877
magnesium;dinitrate;hexahydrate
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
13446-18-9
H12MgN2O12
256.40
MFCD00149779
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
202877
magnesium;dinitrate;hexahydrate
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 330.0°C
Melting Point 89.0°C
Color White
pH 5.0 to 8.2 (5% soln. at 25°C)
Physical Form Adhering Crystalline Powder
Quantity 500 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula Mg(NO3)2·6H2O
Flash Point 93.3°C
Merck Index 15, 5738
Solubility Information Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3
Formula Weight 256.4
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic Bottle
Density 1.63
Chemical Name or Material Magnesium nitrate hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'