missing translation for 'onlineSavingsMsg'
Learn More

Magnesium methoxide, 7-8% in methanol, Thermo Scientific Chemicals

Catalog Number p-7051098
Click to view available options
Quantity:
250 g
1 kg
2 kg
This item is not returnable. View return policy
109-88-6
MFCD00053671
This item is not returnable. View return policy
109-88-6
MFCD00053671

MgO films

The use of magnesium methoxide for the deprotection of alkyl esters is described. This mild reagent provides a good method to cleave esters efficiently and more importantly, allows for effective differentiation between two different esters. Dimethyl carbonate is synthesized directly from carbon dioxide and methanol in the presence of magnesium methoxide. 20 mol% DEA additions to magnesium methoxide showed enhanced stability such that a time-dependent change in the sol was not observed in air. Also enhanced the crystallization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.815 g/mL
Flash Point 14°C (57°F)
Odor Alcohol-like
Refractive Index 1.338
Quantity 250 g
UN Number UN1230
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 86.38
Concentration or Composition (by Analyte or Components) 7 to 8% in methanol
Physical Form Liquid
Chemical Name or Material Magnesium methoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'