missing translation for 'onlineSavingsMsg'
Learn More

Magnesium iodide, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog Number 11317867
Click to view available options
Quantity:
1 g
10 g
10377-58-9
I2Mg
278.11
MFCD00049485
BLQJIBCZHWBKSL-UHFFFAOYSA-L
magnesium(2+) diiodide
[Mg++].[I-].[I-]
10377-58-9
I2Mg
278.11
MFCD00049485
BLQJIBCZHWBKSL-UHFFFAOYSA-L
magnesium(2+) diiodide
[Mg++].[I-].[I-]

Light Sensitive and Hygroscopic

Magnesium iodide is used as reagents and intermediates in pharmaceuticals. It is useful in Baylis-Hillman reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 637°C (decomposition)
Color White
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.996% (Metals basis)
Solubility Information Soluble in water(1480g/L at 18 C)
Formula Weight 278.12
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 4.43g/cm3 at 20°C
Chemical Name or Material Magnesium iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'