missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, Thermo Scientific Chemicals

Catalog Number 12635157
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
7782-75-4
H7MgO7P
174.33
MFCD00150390
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
16204530
magnesium;hydrogen phosphate;trihydrate
O.O.O.[Mg++].OP([O-])([O-])=O
This item is not returnable. View return policy
7782-75-4
H7MgO7P
174.33
MFCD00150390
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
16204530
magnesium;hydrogen phosphate;trihydrate
O.O.O.[Mg++].OP([O-])([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 kg
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula MgHPO4·3H2O
Solubility Information (0.1 M in 5 M HCl) Clear colorless
Formula Weight 174.34
Percent Purity 98%
Grade Biochemical
Packaging Plastic bottle
Specific Gravity 2.13
Density 2.1300g/mL
Chemical Name or Material Magnesium hydrogen phosphate trihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'