missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium hexafluorosilicate hydrate, Thermo Scientific Chemicals
108.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
Description
It is employed as an antiplaque agents and oral care agents. Also used for mothproofing textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 120°C (decomposition) |
Physical Form | Powder |
Quantity | 2 kg |
UN Number | UN2853 |
Solubility Information | Soluble in water. |
Formula Weight | 166.39 (Anhydrous) |
Density | 1.788 g/mL |
Chemical Name or Material | Magnesium hexafluorosilicate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'