missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium citrate hydrate, Mg ca 16% (dry wt.), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
17 kg
Description
Used as a laxative or dietary supplement
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 17 kg |
Merck Index | 14,5663 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 451.13 (Anhydrous) |
Concentration or Composition (by Analyte or Components) | Mg ≈ 16% (dry wt.) |
Chemical Name or Material | Magnesium citrate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'