missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Melting Point | 117°C |
Color | White |
pH | 5 to 6.5 |
Physical Form | Powder/Solid |
Quantity | 1 kg |
Formula Weight | 203.31g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'