missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, >98%, Alfa Aesar™

Brand:  Alfa Aesar 012288.36

Code : NEW

Additional Details : CAS Number : 7791-18-6 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 11341639

  • £13.10 / 500g


EU Stock 14
Add to basket

Specifications

Chemical Identifiers

Magnesium chloride hexahydrate
Flakes
MgCl2·6H2O
MFCD00149781
7791-18-6
>98%
203.30 (95.22anhydrous)

Specifications

Magnesium chloride hexahydrate
Flakes
MgCl2·6H2O
203.30 (95.22anhydrous)
1.57g/cm3 at 20°C
White
7791-18-6
>98%
White flakes
MFCD00149781
Odorless
500g
Videos
SDS