missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium chloride hexahydrate, 98+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 117.0°C |
Quantity | 5 kg |
Assay Percent Range | 98.0 to 101% (Complexometry) |
Linear Formula | MgCl2·6H2O |
Merck Index | 15,5726 |
Solubility Information | Solubility in water: 2350g/L (20°C). Other solubilities: soluble in alcohol |
Formula Weight | 203.31 |
Percent Purity | ≥98% |
Grade | Extra Pure |
Chemical Name or Material | Magnesium chloride hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'