missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, anhydrous, 99%, Alfa Aesar™

For Use As a Catalyst, Flocculating Agent, Various Magnesium Cements

Brand:  Alfa Aesar 012315.A7

Code : NEW

Additional Details : CAS Number : 7786-30-3 Weight : 5.00000kg

Quantity 5kg

 View more versions of this product

Product Code. 11391679

  • £96.00 / 5kg


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Chemical Identifiers

Magnesium Chloride, Anhydrous
Crystals
MgCl2
MFCD00011106
7786-30-3
99%
95.22

Specifications

Magnesium Chloride, Anhydrous
Crystals
MgCl2
95.22
2.32g/cm3 at 20°C
White to yellow
7786-30-3
99%
White to yellow crystals
MFCD00011106
Odorless
5kg
Videos
SDS