missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium carbonate hydroxide tetrahydrate, White powder, MgO approx.40-43.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Magnesium carbonate hydroxide tetrahydrate is used as a mixture with huntite and as filler in polymers. It acts as a flame retardant and fire retardant additive for polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | MgO approx.40-43.5% |
Solubility Information | Slightly soluble in water. |
Formula Weight | 467.67 (395.58 Anhydrous) |
Grade | Reagent |
Odor | Odorless |
Chemical Name or Material | Magnesium carbonate hydroxide tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'