missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number 10703591
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
13446-53-2
Br2H12MgO6
292.20
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
dibromomagnesium hexahydrate
O.O.O.O.O.O.Br[Mg]Br
This item is not returnable. View return policy
13446-53-2
Br2H12MgO6
292.20
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
dibromomagnesium hexahydrate
O.O.O.O.O.O.Br[Mg]Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
pH 6.5 to 8 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99+%
Linear Formula MgBr2·6H2O
Fieser 13,259; 15,194; 16,199
Merck Index 14, 5659
Formula Weight 292.21
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Magnesium bromide hexahydrate, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium bromide hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'