missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080046
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
13446-53-2
Br2H12MgO6
292.20
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
O.O.O.O.O.O.Br[Mg]Br
This item is not returnable. View return policy
13446-53-2
Br2H12MgO6
292.20
MFCD00149780
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
17749097
O.O.O.O.O.O.Br[Mg]Br

Magnesium bromide hexahydrate is used as flame retardant. Used as source of magnesium ion and a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172°C
Color Colorless
Physical Form Crystalline
Quantity 100 g
Linear Formula MgBr2·6H2O
Solubility Information Soluble in water, somewhat soluble in alcohol
IUPAC Name dibromomagnesium hexahydrate
Formula Weight 292.22 (184.13 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.00 g/mL
Chemical Name or Material Magnesium bromide hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'