missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Magnesium Aluminum Zinc wire, 1.59mm (0.063 in.) dia., 90cm (35 in.) long

Brand:  Alfa Aesar™ 014069.RN

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 5 x 90cm

 View more versions of this product

Product Code. 11397677

  • £31.00 / Pack of 5


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

Magnesium Aluminum Zinc wire
1.59mm (0.063 in.) dia., 90cm (35 in.) long
MFCD01074901

Specifications

Mg:Al:Zn; 93:6:1 wt%
≈3.1g/90cm
1.59mm (0.063 in.) dia., 90cm (35 in.) long
MFCD01074901
Magnesium Aluminum Zinc wire
5 x 90cm
Wire
Odorless
Videos
SDS