missing translation for 'onlineSavingsMsg'
Learn More

Magnesium aluminum oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049800
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
12068-51-8
Al2MgO4
142.26
MFCD00049475
UAMZXLIURMNTHD-UHFFFAOYSA-N
alumina magnesia, cernel i, dialuminum magnesium tetraoxide, magnesium aluminate mgal2o4, dialuminum;magnesium;oxygen 2-, magnesium aluminate mg alo2 2, aluminum magnesium oxide, aluminum magnesium oxide mgal2o4, magnesium aluminum oxide mgal2o4, aluminate alo21-, magnesium 2:1
This item is not returnable. View return policy
12068-51-8
Al2MgO4
142.26
MFCD00049475
UAMZXLIURMNTHD-UHFFFAOYSA-N
alumina magnesia, cernel i, dialuminum magnesium tetraoxide, magnesium aluminate mgal2o4, dialuminum;magnesium;oxygen 2-, magnesium aluminate mg alo2 2, aluminum magnesium oxide, aluminum magnesium oxide mgal2o4, magnesium aluminum oxide mgal2o4, aluminate alo21-, magnesium 2:1

Magnesium aluminum oxide is used in chemical research, chemicals, pharmaceutical research, pharmaceutical intermediates reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 100 g
Solubility Information Not soluble in water.
Formula Weight 142.26
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Magnesium aluminum oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'