Learn More
Thermo Scientific Alfa Aesar Magnesium aluminum oxide, 99.9% (metals basis excluding Ca), Ca <25ppm
Brand: Thermo Scientific Alfa Aesar 036388.22
Magnesium aluminum oxide, 99.9% (metals basis excluding Ca), Ca <25ppm
Code : NEW
Additional Details : CAS Number : 12068-51-8 Weight : 0.10000kg