missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate tetrahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog Number p-3208497
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
16674-78-5
C4H14MgO8
214.45
MFCD00149214
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
134717
magnesium(2+) diacetate tetrahydrate
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
16674-78-5
C4H14MgO8
214.45
MFCD00149214
XKPKPGCRSHFTKM-UHFFFAOYSA-L
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
134717
magnesium(2+) diacetate tetrahydrate
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Used as a source of magnesium in biological reactions and as a source of acetate ion in chemical transformations. Acts as an emission control agent in coal combustion process to reduce acid rain. It is involved in glass formation process via the thermal decomposition of solids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 98.0-102.0%
Beilstein 3730605
Solubility Information Soluble in water.
Formula Weight 214.45 (142.39 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystals
Density 1.454 g/cm3 at 20°C
Chemical Name or Material Magnesium acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'