Learn More
Thermo Scientific™ MagnaBind™ Amine Derivatized Beads
Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.
Brand: Thermo Scientific™ 21352
118.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : PN
Additional Details : CAS Number : 79-06-1 Weight : 0.80000kg
Description
Thermo Scientific™ MagnaBind Magnetic Beads are available with streptavidin, Protein A, Protein G, GAM IgG, GAR IgG and other ligands for use in small-scale benchtop magnetic affinity protocols.
MagnaBind Magnetic Beads enable small-scale affinity purification and cell-separation without columns or centrifugation. The beads are silanized iron oxide magnetic particles activated with various affinity ligands. After a binding step in an affinity purification procedure, the magnetic particles are easily and rapidly collected by placing the microcentrifuge tube or reaction vessel next to an appropriate rare-earth magnet. MagnaBind Beads respond rapidly to MagnaBind Magnets. The particles can be easily dispersed and regathered multiple times (i.e., they will not irreversibly aggregate) because they do not have any magnetic memory.MagnaBind Beads are available pre-coated with Protein A, Protein G, streptavidin, anti-mouse or anti-rabbit antibodies. Also available are beads activated with either amine or carboxyl groups as targets for customized covalent immobilization other proteins or ligands.
Highlights:
- Available pre-coated with popular affinity ligands or derivatized for covalent attachment of proteins and other specific ligands
- Beads do not irreversibly aggregate because they have no magnetic memory; collect and disperse the beads multiple times if needed
- Most separations require a short five- to 10-minute benchtop procedure
Properties of underivatized MagnaBind Beads:
- Composition: silanized iron oxide
- Magnetization: 25 to 35 EMU/g
- Type of Magnetization: superparamagnetic (no magnetic memory)
- Surface Area: greater than 100m2/g
- Bead Size: 1 to 4μm diameter
- Settling Rate: 4% in 30 minutes
- Effective Density: 2.5g/mL
- Number of Beads: 1 x 108 beads per milliliter
- pH Stability: aqueous solution, above pH 4.0
- Concentration: 5mg/mL
Requires:
To remove MagnaBind Beads from suspension, an external magnetic field is used; see the ordering table for a selection of magnets.
Recommended for:
Cell sorting using either positive or negative selection; Protein purification or immunoassays using either direct or indirect methods; Immunoprecipiation
Specifications
Upon receipt store product at 4°C. Do not freeze product. Product is shipped at ambient temperature. | |
Brown | |
Amine Derivatized Bead | |
Amine |
MagnaBind Amine Derivatized Beads | |
Affinity Purification | |
MagnaBind™ | |
1 to 4 μm |
For Research Use Only. Not for use in diagnostic procedures.