Learn More
Thermo Scientific™ MagJET Genomic DNA Kit
Purify genomic DNA from cell culture, tissue samples, bacteria, and yeast with a magnetic bead separation method amenable to high-throughput workflows.
Brand:  Thermo Scientific™ K2722
Code : 88
Additional Details : Weight : 1.50000kg
 View more versions of this product
Product Code. 13585170
  • £763.87 / Pack of 4
Estimated Shipment
03-05-2018
Add to basket
Description
Description
Highlights
  • Versatile -suitable for gDNA purification from variety of biological samples, including gDNA from eukaryotic cells, animal tissues, yeast and bacteria, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles
  • Efficient -high yields of high molecular weight genomic DNA
  • High quality -isolates high integrity gDNA, free of inhibitors and RNA contamination
  • Flexible -easily adjustable for high throughput automation platforms and manual processing
Specifications
Specifications
4 x 96 Preps
Ambient
Videos