missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, 99+%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3737748
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-38-3
C8H10
106.17
MFCD00008536
IVSZLXZYQVIEFR-UHFFFAOYSA-N
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
7929
CHEBI:28488
1,3-xylene
CC1=CC(C)=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-38-3
C8H10
106.17
MFCD00008536
IVSZLXZYQVIEFR-UHFFFAOYSA-N
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
7929
CHEBI:28488
1,3-xylene
CC1=CC(C)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -48.0°C
CAS Min % 99.0
Color Colorless
CAS Max % 100.0
Density 0.8640g/mL
Boiling Point 139.0°C
Quantity 250 mL
Flash Point 25°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4960 to 1.4980
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.864
Linear Formula C6H4(CH3)2
Viscosity 0.62 mPa.s (20°C)
Beilstein 05, 370
Water 0.1% max.
Merck Index 15, 10278
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 106.17
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material m-Xylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • m-Xylene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'