missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ m-Tolylacetonitrile, 98%

Brand:  Alfa Aesar™ A10558.06

Code : NEW

Additional Details : CAS Number : 2947-60-6 Weight : 0.00500kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11483194

  • £12.40 / 5g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

m-Tolylacetonitrile
C9H9N
MFCD00001914
1099644
2947-60-6
131.18
UN3276
3-Methylbenzyl cyanide; 3-Methylphenylacetonitrile

Specifications

m-Tolylacetonitrile
98%
131.18
Almond-like
1099644
240° to 241°C
5g
2947-60-6
C9H9N
MFCD00001914
UN3276
3-Methylbenzyl cyanide; 3-Methylphenylacetonitrile
>110°C (230°F)
Videos
SDS