Learn More
Alfa Aesar™ m-Tolylacetonitrile, 98%
Brand: Alfa Aesar™ A10558.22
Code : NEW
Additional Details : CAS Number : 2947-60-6 Weight : 0.10000kg Transport : UN number : 3276 Chem class : 1 Pack group : III
Quantity | 100g |
---|
Chemical Identifiers
m-Tolylacetonitrile | |
C9H9N | |
MFCD00001914 | |
1099644 |
2947-60-6 | |
131.18 | |
UN3276 | |
3-Methylbenzyl cyanide; 3-Methylphenylacetonitrile |
Specifications
m-Tolylacetonitrile | |
98% | |
131.18 | |
Almond-like | |
1099644 | |
240° to 241°C | |
100g |
2947-60-6 | |
C9H9N | |
MFCD00001914 | |
UN3276 | |
3-Methylbenzyl cyanide; 3-Methylphenylacetonitrile | |
>110°C (230°F) |