missing translation for 'onlineSavingsMsg'
Learn More

m-Tolunitrile, 99%, Thermo Scientific Chemicals

Catalog Number p-3758828
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
620-22-4
C8H7N
117.15
MFCD00001808
BOHCMQZJWOGWTA-UHFFFAOYSA-N
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
12104
3-methylbenzonitrile
CC1=CC=CC(=C1)C#N
This item is not returnable. View return policy
620-22-4
C8H7N
117.15
MFCD00001808
BOHCMQZJWOGWTA-UHFFFAOYSA-N
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
12104
3-methylbenzonitrile
CC1=CC=CC(=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -23°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 99°C to 101°C (20.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4CN
Refractive Index 1.5246 to 1.5266
Quantity 5 g
Beilstein 09, 477
Specific Gravity 0.97
Solubility Information Solubility in water: .. Other solubilities: solunle in methanol,ethanol,dioxane
Formula Weight 117.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Tolunitrile
Show More Show Less
compliance-icons
Product Identifier
  • m-Tolunitrile
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'