missing translation for 'onlineSavingsMsg'
Learn More

m-Hydroxycinnamic acid, 99%, predominantly trans, Thermo Scientific Chemicals

Catalog Number 10061090
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
14755-02-3
C9H8O3
164.16
MFCD00004390
KKSDGJDHHZEWEP-SNAWJCMRSA-N
3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
637541
CHEBI:32357
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC(=C1)O)C=CC(=O)O
This item is not returnable. View return policy
14755-02-3
C9H8O3
164.16
MFCD00004390
KKSDGJDHHZEWEP-SNAWJCMRSA-N
3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
637541
CHEBI:32357
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC(=C1)O)C=CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 189°C to 192°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula HOC6H4CH=CHCO2H
Quantity 25 g
Beilstein 10, 294
Formula Weight 164.16
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material m-Hydroxycinnamic acid, Predominantly trans
Show More Show Less
compliance-icons
Product Identifier
  • m-Hydroxycinnamic acid, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'