missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts M.faeni Serological Double Gel Diffusion Assay
Microgen Bioproducts M.faeni- A serological double gel diffusion assay for detection of precipitating antibodies to the thermophilic actinomycete Micropolyspora faeni.
Brand: Novacyt - Microgen Bioproducts FSK3
Additional Details : Weight : 0.50000kg
Description
- Double diffusion assay for the detection of precipitating antibodies to thermophilic actinomycetes.
Specifications
6 mL | |
Double gel diffusion (Ouchterlony) | |
Test Kit | |
5 to 10 mL of venous blood |
Liquid | |
Double Gel Diffusion Assay | |
EC/REP label on kit box, Blank CE Carton label |