missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-CSFR/CD115 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | M-CSFR/CD115 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115 Ile20-Glu512 Accession # CAA27300 |
Gene ID (Entrez) | 1436 |
Gene Accession No. | CAA27300 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
M-CSFR/CD115 Polyclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
M-CSFR/CD115 | |
1436 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human M-CSF R/CD115 in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot, Flow Cytometry, CyTOF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115 Ile20-Glu512 Accession # CAA27300 | |
CAA27300 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only