missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals m-Cresol Purple, pure, indicator grade

Catalog Number 10013030
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2303-01-7
C21H18O5S
382.43
MFCD00005871
OLQIKGSZDTXODA-UHFFFAOYSA-N
73030
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C
This item is not returnable. View return policy
2303-01-7
C21H18O5S
382.43
MFCD00005871
OLQIKGSZDTXODA-UHFFFAOYSA-N
73030
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Formula Weight 382.42
Grade Pure
Packaging Glass bottle
Specific Extinction 900 min. (578nm, in buffer)
Visual Transition Interval from pH 7.4 (yellow) to pH 9.0 (purple)
Color Green
Quantity 10 g
Physical Form Fine Crystalline Powder
Chemical Name or Material m-Cresol Purple

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'