missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, 99%, Thermo Scientific Chemicals

Catalog Number 11419612
27.27 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.


Click to view available options
Quantity:
250 g
800 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
108-39-4
C7H8O
108.14
MFCD00002302
RLSSMJSEOOYNOY-UHFFFAOYSA-N
3-Methylphenol
342
CHEBI:17231
3-methylphenol
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
108-39-4
C7H8O
108.14
MFCD00002302
RLSSMJSEOOYNOY-UHFFFAOYSA-N
3-Methylphenol
342
CHEBI:17231
3-methylphenol

m-Cresol is employed in the preparation of synthetic resins like phenol-formaldehyde resins, disinfectants, and photographic developers. It is used as industrial solvent for dissolving polymers such as polyaniline. It acts as a precursor to vitamin E and antiseptic amylmetacresol. It is used as insuline preservatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Cresol is employed in the preparation of synthetic resins like phenol-formaldehyde resins, disinfectants, and photographic developers. It is used as industrial solvent for dissolving polymers such as polyaniline. It acts as a precursor to vitamin E and antiseptic amylmetacresol. It is used as insuline preservatives.

Solubility
Miscible with water, diethylether, chloroform and ethanol.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, aluminum and aluminum alloys.
TRUSTED_SUSTAINABILITY
Melting Point 11°C to 12°C
Boiling Point 202°C to 204°C
UN Number UN2076
Beilstein 506719
Merck Index 14,2579
Solubility Information Miscible with water,diethylether,chloroform and ethanol.
SMILES CC1=CC(=CC=C1)O
Formula Weight 108.14
Percent Purity 99%
Odor Phenol-like
Sensitivity Light sensitive
Density 1.034
pH 5
Quantity 250 g
Flash Point 86°C (186°F)
Chemical Name or Material m-Cresol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'