missing translation for 'onlineSavingsMsg'
Learn More

m-Anisidine, 99%, Thermo Scientific Chemicals

Catalog Number 10606482
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
This item is not returnable. View return policy
536-90-3
C7H9NO
123.15
MFCD00007783
NCBZRJODKRCREW-UHFFFAOYSA-N
m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i
10824
3-methoxyaniline
COC1=CC=CC(=C1)N
This item is not returnable. View return policy
536-90-3
C7H9NO
123.15
MFCD00007783
NCBZRJODKRCREW-UHFFFAOYSA-N
m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i
10824
3-methoxyaniline
COC1=CC=CC(=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 1.1020g/mL
Boiling Point 250°C to 251°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NH2
Refractive Index 1.5800 to 1.582
Quantity 100 mL
Beilstein 13,404
Merck Index 14,667
Specific Gravity 1.102
Solubility Information Solubility in water: 18g/L (20°C). Other solubilities: soluble in acids,soluble in alcohol,diethyl ether,acetone and,benzene
Formula Weight 123.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Anisidine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'