Learn More
Merck Millipore M Air T Cassette Prefilled with Sabouraud Dextrose Agar Media
For pharmaceutical products analysis and environmental analysis
Brand:  Merck Millipore ATSMSDD60
Code : NEW
Additional Details : Weight : 0.80000kg
Product Code. 11963916
  • £365.00 / Pack of 60
Estimated Shipment
17-03-2021
Add to basket
Description
Description
A lightweight air tester featuring a unique cassette design for determining the presence of viable, airborne microorganisms in critical areas. The system impacts microorganisms onto an agar surface, which is the referenced method for analysing microbial air contamination (USP 24, p. 2099).Air monitoring systems must be calibrated in order to conform to Good Manufacturing Practices. Using the M Air T calibration kit. The kit contains everything you need: flow meter, flow rate correction table, instruction manual, calibration sieve, and calibration certificate.
  • Ready-to-use cassettes, pre-filled with medium, gamma sterilised, double sleeve packaged, certificate included
  • Easy-to-use, compact, lightweight sampler, programmable, extended life battery, delay timer to minimise risk of secondary contamination
  • USP and EP compliance certificate
  • Simple colony counting through integrated grid
  • Fast and reliable results with maximum recovery; large agar amount and unique micro-perforated sieve allow air sampling of up to 1m3
  • Reproducible results; unit-to-unit consistency of agar surface shape
Specifications
Specifications
1Pack
For pharmaceutical products analysis and environmental analysis
M Air T Cassette prefilled with Sabouraud Dextrose Agar Media
5.6 ± 0.2pH