missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysine Hydroxylase 2/PLOD2 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | Lysine Hydroxylase 2/PLOD2 |
---|---|
Clone | 408105 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Lysine Hydroxylase 2/PLOD2 Monoclonal specifically detects Lysine Hydroxylase 2/PLOD2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Lysine Hydroxylase 2/PLOD2 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
NP_891988 | |
5352 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Lysine Hydroxylase 2/PLOD2 Ala26-Pro758 Accession # NP_891988 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Lysine Hydroxylase 2/PLOD2 in direct ELISAs and Western blots. |
408105 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
2-oxoglutarate 5-dioxygenase (lysine hydroxylase) 2, 2-oxoglutarate 5-dioxygenase 2, LH2, PLOD2, procollagen-lysine, telopeptide lysyl hydroxylase, TLH | |
PLOD2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only