missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lys-Bradykinin Acetate Salt

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J61171.MB

Code : NEW

Additional Details : CAS Number : 342-10-9 Weight : 0.00025kg

 View more versions of this product

Product Code. 15493789

  • £211.80 / 25mg


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Specifications

1188.40
25mg
Powder
RUO
Kallidin
HPLC
Endogenous bradykinin receptor agonist
Lys-Arg-Pro-Pro-Gly- Phe-Ser-Pro-Phe-Arg
-30° to -10°C
Lys-Bradykinin acetate salt
Videos
SDS

For Research Use Only