Learn More
Sartorius™ Lyphan Indicator Strips
Brand:  Sartorius™ FT-2-00603-0405
UNSPSC : 41000000
Code : NEW
Additional Details : Weight : 0.06000kg
Product Code. 10128073
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications Description & Specifications
Specifications
For accurate determination of pH in water samples and for adjusting the pH of a sample during analysis
200/Pk.
Lyphan L 662 Indicator Strip
Strip
Lyphan L 662
3.9 to 5.4