Learn More
Lymphotoxin beta/TNFSF3 Mouse anti-Human, Clone: 135105, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1684
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448825
  • £390.72 / 500µg
Estimated Shipment date
from Supplier 07-08-2020
Description
Description
Lymphotoxin beta/TNFSF3 Monoclonal antibody MAB1684 Specifically detects Lymphotoxin beta/TNFSF3 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Lymphotoxin beta/TNFSF3
135105
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
LTB, lymphotoxin beta (TNF superfamily, member 3), lymphotoxin-beta, p33, TNFCTNF-C, TNFSF3, TNFSF3LT-beta, Tumor necrosis factor C, Tumor necrosis factor ligand superfamily member 3
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Lymphotoxin alpha beta heterotrimers in direct ELISAs and Western blots. This antibody detects an epitope in the Lymphotoxin beta subunit. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Lymphotoxin alpha , rhAPRIL, rhBAFF, rhTNF-alpha , rhTWEAK, rhEDA-A2, rhEDA, rhTRAIL, rhVEGI, rhTRANCE, rhFas Ligand, or rhOX40 Ligand.
Human
Western Blot
LYOPH
Western Blot 1 ug/mL
Purified
LTB
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Lymphotoxin alpha/beta
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4050
Reconstitute at 0.5 mg/mL in sterile PBS.